Learn More
Med Vet International Miltex Meisterhand Russian Tissue Forceps, 10in, German, Mh6-145
Catalog No. 502832255
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Russian Tissue Forceps. Russian Tissue Forceps are thumb tissue forceps specially crafted to grasp dense tissues and stabilize them during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 10in, MH6-145
Catalog No. 50-283-2255 Supplier Med Vet International Supplier No. MILMH6145
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Russian Tissue Forceps. Russian Tissue Forceps are thumb tissue forceps specially crafted to grasp dense tissues and stabilize them during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 10in, MH6-145