Learn More
Research Products International Corp Ergonomic Forceps, Serrated, 3mm Tip, 7 Inches Long
SDP
Catalog No. 50212612
Encompass
Ergonomic forceps are specially designed with a wide paddle handle and reduced tension. Lightweight forceps reduce hand fatigue and are effortless to operate. Forceps are easy to manage during procedures involving tissue dissection, embedding, preparation, and much more.
Catalog No. 50-212-612 Supplier Research Products International Corp Supplier No. 248833
Ergonomic forceps are specially designed with a wide paddle handle and reduced tension. Lightweight forceps reduce hand fatigue and are effortless to operate. Forceps are easy to manage during procedures involving tissue dissection, embedding, preparation, and much more.