Learn More
Fine Science Tools Tissue Forceps, Slim 1 x 2 Teeth, Stainless Steel, 10 cm
SDP
Catalog No. NC9657173
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 1.7 x 1.3mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9657173 Supplier Fine Science Tools Supplier No. 1102310
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 1.7 x 1.3mm/ Alloy / Material: Stainless Steel/ Length: 10cm