Learn More
Med Vet International Miltex Vantage Splinter Forceps, 3.5in
Catalog No. 502522269
Encompass_Preferred
The Miltex Vantage V96-300 Splinter Forceps are made of mid-grade surgical steel and have a length of 3.5in. Splinter Forceps are a fine-tipped forceps, used in first aid, to remove tiny foreign bodies from tissues, and may be used to handle sutures.
Catalog No. 50-252-2269 Supplier Med Vet International Supplier No. MILV96300
The Miltex Vantage V96-300 Splinter Forceps are made of mid-grade surgical steel and have a length of 3.5in. Splinter Forceps are a fine-tipped forceps, used in first aid, to remove tiny foreign bodies from tissues, and may be used to handle sutures.