Learn More
BURKLE INC Forceps, stainless steel, blunt, straight, 6.30 in. (160 mm)
Catalog No. 503081266
Encompass_Preferred
These stainless steel forceps are suitable for precise laboratory tasks. The forceps have a straight shape with blunt ends that are fluted on the inside. This makes it possible to pick up different materials, tissues or samples accurately. The laboratory tweezers are made of stainless steel and feature a fluted handle.
Catalog No. 50-308-1266 Supplier BURKLE INC Supplier No. 53860304
These stainless steel forceps are suitable for precise laboratory tasks. The forceps have a straight shape with blunt ends that are fluted on the inside. This makes it possible to pick up different materials, tissues or samples accurately. The laboratory tweezers are made of stainless steel and feature a fluted handle.