Learn More
BURKLE INC Forceps, stainless steel, sharp, straight, 4.13 in. (105 mm)
Catalog No. 503081248
Encompass_Preferred
The sturdy stainless steel forceps are ideal for use in laboratories. Pointed ends with a fluted internal structure ensure reliable handling of different samples. The fluted handle offers a good grip. The laboratory tweezers are suitable for use in precise work in chemistry, the pharmaceutical industry, cosmetics and food & beverage.
Catalog No. 50-308-1248 Supplier BURKLE INC Supplier No. 53860200
The sturdy stainless steel forceps are ideal for use in laboratories. Pointed ends with a fluted internal structure ensure reliable handling of different samples. The fluted handle offers a good grip. The laboratory tweezers are suitable for use in precise work in chemistry, the pharmaceutical industry, cosmetics and food & beverage.