Learn More
Fine Science Tools Standard Pattern Forceps, Stainless Steel, 30 cm
SDP
Catalog No. NC9927963
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 4 x 2.25mm/ Alloy / Material: Stainless Steel/ Length: 30cm
Catalog No. NC9927963 Supplier Fine Science Tools Supplier No. 1100030
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 4 x 2.25mm/ Alloy / Material: Stainless Steel/ Length: 30cm