Learn More
Andwin Scientific FORCEPS STER DISP PS PK100
SDP
Catalog No. NC1474836
Encompass_Preferred
Laboratory Forceps With blunt tip. Disposable polystyrene. Sterile. Color: blue. Individual peel pack. 100/pack
Catalog No. NC1474836 Supplier Andwin Scientific Supplier No. 12576934PK
Laboratory Forceps With blunt tip. Disposable polystyrene. Sterile. Color: blue. Individual peel pack. 100/pack