Learn More
Fine Science Tools Standard Pattern Forceps, Stainless Steel, 12 cm
SDP
Catalog No. NC9457310
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 2.5 x 1.35mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9457310 Supplier Fine Science Tools Supplier No. 1100012
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 2.5 x 1.35mm/ Alloy / Material: Stainless Steel/ Length: 12cm