Learn More
Fine Science Tools Taylor Forceps, Stainless Steel, Straight, 17 cm
SDP
Supplier:  Fine Science Tools 1101617
Encompass
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 1.5 x 1mm/ Alloy / Material: Stainless Steel/ Length: 17cm
Catalog No. NC9057875