Learn More
Med Vet International Miltex Swiss-Jeweler Splinter Forceps, 4.375in., Non-Magnetic, Super Fine Tips
Catalog No. 502830428
Encompass_Preferred
The Miltex 17-305 Swiss-jeweler splinter forceps are made of OR grade German stainless steel and measure 4.375" in length. They have a non-locking thumb handle and tips that are straight and super fine. Non-magnetic.
Catalog No. 50-283-0428 Supplier Med Vet International Supplier No. MIL17305
The Miltex 17-305 Swiss-jeweler splinter forceps are made of OR grade German stainless steel and measure 4.375" in length. They have a non-locking thumb handle and tips that are straight and super fine. Non-magnetic.