Learn More
Fine Science Tools FORCEPS TIP DIAMETER 0.2MM
SDP
Catalog No. NC2034560
Encompass
Item No. 00125-11S&T Vessel Dilating Forceps - Angled 10 degreesTip Shape:Angled 10°Alloy / Material:Stainless SteelLength:11cmTip Diameter:0.2mm
Catalog No. NC2034560 Supplier Fine Science Tools Supplier No. 0012511
Item No. 00125-11S&T Vessel Dilating Forceps - Angled 10 degreesTip Shape:Angled 10°Alloy / Material:Stainless SteelLength:11cmTip Diameter:0.2mm