Learn More
Med Vet International Tissue Forceps, 5.75", Standard Pattern, Serrated Handles, 3x4 Teeth, German Grade, Von Klaus
Catalog No. 503046160
Encompass_Preferred
5 3/4in. tissue forceps have a standard pattern, serrated handles, and 3x4 teeth. Tissue forceps are great to use for holding and manipulating tissues durring multiple surgical procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6160 Supplier Med Vet International Supplier No. VK1061114
5 3/4in. tissue forceps have a standard pattern, serrated handles, and 3x4 teeth. Tissue forceps are great to use for holding and manipulating tissues durring multiple surgical procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.