Learn More
Med Vet International Miltex Russian Tissue Forceps, 6in, Serrated
Catalog No. 502522291
Encompass_Preferred
The Miltex 6-142 Russian tissue forceps are made of OR grade German stainless steel and measure 6" in length. They have a non-locking thumb handle and straight, serrated tips.
Catalog No. 50-252-2291 Supplier Med Vet International Supplier No. MIL6142
The Miltex 6-142 Russian tissue forceps are made of OR grade German stainless steel and measure 6" in length. They have a non-locking thumb handle and straight, serrated tips.