Learn More
World Precision Instrument Backhaus Towel Clamp, 13cm, Curved, Locking handle, SS
SDP
Catalog No. 50822701
Encompass
Backhaus Towel Clamps, also known as the Backhaus forceps or towel forceps, are designed for securing and attaching surgical drapes or towels to a patient's skin or other surfaces. The Backhaus Towel Clamp has a scissor-like design with ring handles. These curved, pointed jaws have small, sharp, and inward-facing tips for a secure grip when the instrument is closed. A ratchet mechanism lets you securely lock them in a closed position to maintain a firm grip on the drapes. Our Backhaus Towel Clamps are available in 9 cm (3.5") or 13 cm (5.25"). Choose the size based on the thickness of the material to be clamped and the area of application. These surgical forceps are manufactured with surgical grade stainless steel or premium German steel, both of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.
Catalog No. 50-822-701 Supplier World Precision Instrument Supplier No. 501700
Backhaus Towel Clamps, also known as the Backhaus forceps or towel forceps, are designed for securing and attaching surgical drapes or towels to a patient's skin or other surfaces. The Backhaus Towel Clamp has a scissor-like design with ring handles. These curved, pointed jaws have small, sharp, and inward-facing tips for a secure grip when the instrument is closed. A ratchet mechanism lets you securely lock them in a closed position to maintain a firm grip on the drapes. Our Backhaus Towel Clamps are available in 9 cm (3.5") or 13 cm (5.25"). Choose the size based on the thickness of the material to be clamped and the area of application. These surgical forceps are manufactured with surgical grade stainless steel or premium German steel, both of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.