Learn More
Fine Science Tools Tissue Forceps, 1 x 2 Teeth, Stainless Steel, 14.5 cm
SDP
Catalog No. NC9314088
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 2 x 1.5mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm
Catalog No. NC9314088 Supplier Fine Science Tools Supplier No. 1102114
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 2 x 1.5mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm