Learn More
World Precision Instrument Forceps with Tying Platform, 8.5cm, Straight, Titanium
SDP
Catalog No. 502538305
Encompass
  • 8.5cm (3.3") Long
  • Tips: 12mm, Straight
  • 4.5mm Tying platform
  • Titanium
Catalog No. 50-253-8305 Supplier World Precision Instrument Supplier No. WP5000
  • 8.5cm (3.3") Long
  • Tips: 12mm, Straight
  • 4.5mm Tying platform
  • Titanium