Learn More
Med Vet International Desmarres Chalazion Forceps, 3.5", Medium, German Grade, Von Klaus
Catalog No. 502091934
Encompass_Preferred
3.5in. Desmarres Chalazion forceps are German grade surgical instrument use in ophthalmic procedures. They have a medium inside ring that is 23.0mm x 12.0mm. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1934 Supplier Med Vet International Supplier No. VK1605502
3.5in. Desmarres Chalazion forceps are German grade surgical instrument use in ophthalmic procedures. They have a medium inside ring that is 23.0mm x 12.0mm. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.