Learn More
Med Vet International 5 - 5.5-in Economy Mosquito Forceps, Curved
Catalog No. 502224020
Encompass_Preferred
Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries.
Catalog No. 50-222-4020 Supplier Med Vet International Supplier No. 4511
Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries.