Learn More
Med Vet International 4.75-in (12.0cm) Von Klaus Adson-Brown Tissue Forceps, 7x7 Teeth, Straight Tips, German Surgical Steel
Catalog No. 502091923
Encompass_Preferred
Adson-Brown tissue forceps have straight tips and 7x7 teeth. These forceps are great for holding delicate tissues during plastic surgery, they have a wide, flat thumb grasp area. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1923 Supplier Med Vet International Supplier No. VK1062512
Adson-Brown tissue forceps have straight tips and 7x7 teeth. These forceps are great for holding delicate tissues during plastic surgery, they have a wide, flat thumb grasp area. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.