Learn More
Med Vet International Russian Tissue Forceps, 6in, German Grade, Von Klaus
Catalog No. 502091925
Encompass_Preferred
6' Russian tissue forceps is a German grade surgical instrument. They are a medium to heavy forceps used for grasping heavy or thick tissue. Von Klaus is a supplier of high quality German Grade surgical instruments. 6' Russian tissue forceps are made of German grade surgical instrument. They are a medium to heavy forceps used for grasping heavy or thick tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1925 Supplier Med Vet International Supplier No. VK1071515
6' Russian tissue forceps is a German grade surgical instrument. They are a medium to heavy forceps used for grasping heavy or thick tissue. Von Klaus is a supplier of high quality German Grade surgical instruments. 6' Russian tissue forceps are made of German grade surgical instrument. They are a medium to heavy forceps used for grasping heavy or thick tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.