Learn More
Cayman Chemical Forchlorfenuron, Purity: ≥98%, CAS: 68157-60-8, 100 mg
SDP
Catalog No. 502947289
Encompass_Preferred
A plant growth regulator; stimulates fruit growth, as well as increases fruit size and ripening rate, in kiwifruit when applied topically at 40 mg/L; accelerates fruit growth and increases rind thickness in watermelons
Catalog No. 50-294-7289 Supplier Cayman Chemical Supplier No. 32948100 MG
A plant growth regulator; stimulates fruit growth, as well as increases fruit size and ripening rate, in kiwifruit when applied topically at 40 mg/L; accelerates fruit growth and increases rind thickness in watermelons