Learn More
Medchemexpress LLC HY-B1841 500mg Medchemexpress, Forchlorfenuron CAS:68157-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1841500MG
Encompass_Preferred
Medchemexpress, HY-B1841 500mg Forchlorfenuron CAS:68157-60-8 Forchlorfenuron is plant growth regulator and cytokinin; can be used to increase fruit size of fruits, such as kiwi fruit and grapes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2715