Learn More
Waters Corp Waters Foreline Trap, FL20K, Alumina Sorbent, Mass Spectrometry
Catalog No. 50792157 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Foreline Trap FL20K is an essential component designed for use in mass spectrometry systems. It prevents the back-migration of rotary pump oil and oil mist by utilizing an alumina sorbent. This trap effectively ensures clear analytical results by preventing contamination of samples from rotary pump oil and mist.
  • Prevents the back-migration of rotary pump oil and oil mist in mass spectrometry systems.
  • Utilizes an alumina sorbent for effective trapping of oil mist.
  • Contributes to clear analytical results by preventing sample contamination.
  • Recommended for installation as close to the vacuum system as possible with a valved by-pass line due to susceptibility to absorbing water.
Catalog No. 50-792-157 Supplier Waters Corp Supplier No. 6060106
The Foreline Trap FL20K is an essential component designed for use in mass spectrometry systems. It prevents the back-migration of rotary pump oil and oil mist by utilizing an alumina sorbent. This trap effectively ensures clear analytical results by preventing contamination of samples from rotary pump oil and mist.
  • Prevents the back-migration of rotary pump oil and oil mist in mass spectrometry systems.
  • Utilizes an alumina sorbent for effective trapping of oil mist.
  • Contributes to clear analytical results by preventing sample contamination.
  • Recommended for installation as close to the vacuum system as possible with a valved by-pass line due to susceptibility to absorbing water.