Learn More
Electron Microscopy Sciences Mechanical Pump In-line Foreline Traps 3/4"
SDP
Catalog No. 5025993
Encompass
These rechargeable in-line traps effectively block oil backstreaming from mechanical pumps. Any mechanically pumped system, even one with a turbo pump, should have a foreline traps. The trap is placed in the foreline between the roughing pump and the diffusion or turbo pump. It uses a disposable, oxygen-free copper maze absorbent which requires no liquid nitrogen and will not hold water. Typically the maze is replaced every 1-2 years. The seamless body is available in aluminum.
Catalog No. 50-259-93 Supplier Electron Microscopy Sciences Supplier No. 9105015
These rechargeable in-line traps effectively block oil backstreaming from mechanical pumps. Any mechanically pumped system, even one with a turbo pump, should have a foreline traps. The trap is placed in the foreline between the roughing pump and the diffusion or turbo pump. It uses a disposable, oxygen-free copper maze absorbent which requires no liquid nitrogen and will not hold water. Typically the maze is replaced every 1-2 years. The seamless body is available in aluminum.