Learn More
Electron Microscopy Sciences Forensic Field Sampler Stub w/ Adhesive Carbon Tab, 6 mm
SDP
Catalog No. 501927167
Encompass_Preferred
Glass specimen vial has a high-purity certified 12.7 mm SEM pin stub mounted into the plastic cap. It is ideal for collecting forensic evidence with minimum interference and/or contamination from the sampler. The specimen arrives at the lab ready for carbon coating for SEM/EDX investigation. The Forensic Field Sampler can be securely stored in the glass specimen vial and easily shipped with the cap holding the sampler stub in place. Vial is 25 mm OD x 55 mm high. Adhesive carbon tabs secure the evidence material.
Pin length: 6 mm
Catalog No. 50-192-7167 Supplier Electron Microscopy Sciences Supplier No. 7648035
Glass specimen vial has a high-purity certified 12.7 mm SEM pin stub mounted into the plastic cap. It is ideal for collecting forensic evidence with minimum interference and/or contamination from the sampler. The specimen arrives at the lab ready for carbon coating for SEM/EDX investigation. The Forensic Field Sampler can be securely stored in the glass specimen vial and easily shipped with the cap holding the sampler stub in place. Vial is 25 mm OD x 55 mm high. Adhesive carbon tabs secure the evidence material.
Pin length: 6 mm