Learn More
Mopec Forensic Blue Laser Kits
SDP
Catalog No. 22446284 Shop All Mopec Products
Change view
Click to view available options
Watts:
2 W
5 W
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Watts
22-446-284 2 W
22-446-261 5 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-284 Supplier Mopec Supplier No. FS446
Add to Cart
Add to Cart
High-intensity blue laser illumination helps detect forensic evidence by enhancing fluorescence from biological fluids, fingerprints, and trace materials. The system improves visibility of evidence during crime scene investigation and supports clearer observation and documentation.
Performance
  • Perfect for forensic searches
  • More effective at crime scenes as you can see in far greater distances compared to standard forensic light sources
  • Can be used for field crime scene research, laboratory, medicolegal (including coroners and medical examiners), and hotel room inspection
  • Can identify evidence like trace evidence (e.g. hairs, fibers, etc.), gunshot residue, biological fluids, and impressions
  • No maintenance, calibration, or replacement battery needed
  • Can be mounted on a tripod and used underwater
Construction
  • Kit consists of:
  • (1) Forensic blue laser
  • (1) Case
  • (1) Shoulder strap
  • (1) Charger
  • (1) Rechargeable lithium ion battery system
  • (1) Orange laser goggle
  • 445 nm light beam
  • Portable and cord-free
  • Anti-roll head
Specifications
Product Type Forensic Laser Kit
Watts 2 W