Learn More
Electron Microscopy Sciences Forensic Certified SEM Mount 8 mm pin
SDP
Catalog No. 501901507
Encompass_Preferred
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR . These mounts are 12.7 mm x 11 mm.
Catalog No. 50-190-1507 Supplier Electron Microscopy Sciences Supplier No. 7647505
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR . These mounts are 12.7 mm x 11 mm.