Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Chemistry of Document Analysis Kit
GREENER_CHOICESCI_ED

Catalog No. S96639 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96639 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
Catalog No. S96639 Supplier Innovating Science™ Supplier No. IS9003
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

Includes

Teacher's manual and student guide, analysis copy-masters, three different brands of felt-tip markers, chromatography solvent (15 x 30mL), 15 silica gel chromatography sheets, capillary tubes. Enough materials for 15 tests.

Requires

Chromatography chambers (15 x 500mL beakers), 15 test tubes, aluminum foil

Help solve a crime.

The school library's computers have been stolen. Left behind was a ransom note demanding money. Can students determine which marker was used to write the note?

  • Use thin-layer chromatography to separate the ink on the note and ink found in markers tied to possible suspects
  • Review evidence as to whether or not the ransom note could have been written with a particular marker

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For thin layer chromatography
Includes 3 felt tip markers, 15 x 30mL chromatography solvent un1170, 15 silica gel chromatography sheets, capillary tubes, Teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Forensic Document Analysis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.