Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Chemistry of Drug Detection Kit
SCI_ED

Catalog No. s96646 Shop All Aldon Products
Change view
Click to view available options
Forensic Drug Detection Kit; For 15 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S96646 Forensic Drug Detection Kit; For 15 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, which is known to the State of California to cause and reproductive harm.
Catalog No. S96646 Supplier Innovating Science™ Supplier No. IS9010
WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, which is known to the State of California to cause and reproductive harm.

Includes

Teacher's manual and student guide, analysis copy-masters, control acetylsalicylic acid, chili ingredients, ferric nitrate (0.1M solution), Lugol's iodine, 15 microreaction plates. Enough materials for 15 tests.

Requires

Distilled water

Perform a series of chemical tests as a forensic toxicologist.

Everyone who ate chili in the school cafeteria became ill — could someone have tainted it?

  • Students experiment to see if any of the chili ingredients could have been substituted with aspirin, which appears to have been stolen from the office of the school nurse
  • Students test control acetylsalicylic acid, the chemical name of aspirin, in the lab

Specifications

Certifications/Compliance 49 CFR 173.4
Product Type Forensic Drug Detection Kit
Color White
Grade 7 to 14
Age 13+ years
For Use With (Application) For perform a series of chemical tests as a forensic toxicologist
Includes Teacher's manual and student guide, analysis copy-masters, control acetylsalicylic acid, chili ingredients, ferric nitrate (0.1m solution), Lugols iodine, 15 microreaction plates, enough materials for 15 tests
Quantity 1
Requires Distilled water
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.