Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Chemistry of Hair Analysis Kit
GREENER_CHOICESCI_ED

Catalog No. S96643 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96643 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S96643 Supplier Innovating Science™ Supplier No. IS9007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Teacher's manual and student guide, analysis copy-masters, microscope slides, cover slips, deer hair sample, cat hair sample, four human hair samples, 15 forceps, 15 wax pencils

Requires

Light microscopes

See how scientists use hair to solve a crime.

Determine the origin of hair samples from a crime scene and compare to hair samples from four known suspects.

  • Students will discover the differences between human and animal hair as well as those among different types of human hair
  • Enough materials for 15 tests

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Determine the origin of hair samples from a crime scene and compare to hair samples from four known suspects
Includes Teacher's manual and student guide, analysis copy-masters, microscope slides, cover slips, deer hair sample, cat hair sample, four human hair samples, 15 forceps, 15 wax pencils
Quantity 1
Requires Light microscopes
Dimensions (L x W x H) 9.75 x 6.75 x 3 in.
Class Size 15 Lab Groups
Product Type Hair Analysis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.