Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Chemistry of Unknown Substances Kit
GREENER_CHOICESCI_ED

Catalog No. s96642 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96642 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.
Catalog No. S96642 Supplier Innovating Science™ Supplier No. IS9006
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.

Includes

Teacher's manual and student guide, analysis copy-masters, baking powder, baking soda, corn starch, talcum powder, salt, gelatin, two mystery substances, biuret reagent, Lugol's iodine, acetic acid, reaction plates. Enough materials for 15 tests.

Identify unknown substances recovered in the field.

Along with evidence such as fingerprints, hair and fibers, there may be traces of chemicals or powders left behind by the perpetrator. Evidence of this nature is sent to the crime lab for identification. Use observation skills, sense and chemical test results on a series of known and unknown substances to identify mystery substances.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13+ years
For Use With (Application) For forensic test of unknown substances
Includes 30 Reaction plates, 30g baking powder, 30g baking Soda, 30g Corn Starch, 30g Talcum powder, 30g Salt, 30g Gelatin, 30g mystery substance, 30mL Biuret reagent, 30mL dilute Lugol's Iodine, 30mL Acetic Acid, teacher's manual and student study guide
Quantity 1
Dimensions (L x W x H) 12 x 9.25 x 4 in.
Class Size 15 Lab Groups
Product Type Unknown Substances Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.