Learn More
Mopec Forensic Kit
SDP
Catalog No. 22445963 Shop All Mopec Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
22-445-963 1 Kit
1 options
Catalog No. 22-445-963 Supplier Mopec Supplier No. FS224
Add to Cart
Add to Cart
Designed for bloodstain detection, with tablets that dissolve in a prefilled spray bottle for quick and convenient reagent preparation.
  • 1 Bluestar kit box
  • 1 Kit instruction sheet
  • 1 Hermetically sealed bottle with 500 mL (16 oz.) reagent and safety band
  • Includes 3 catalyst tablets.
  • 1 Fine mist atomizer
  • Includes everything needed for locating bloodstains at a crime scene.
  • Provides a complete, easy to use setup for reliable field application.
Specifications
Product Type Forensic Kit
For Use With (Application) Bloodstain Detection
Quantity 1 Kit