Learn More
Motic Swift Line Forensic Microscope Tablet
SCI_ED
Catalog No. S000292 Shop All Motic Swift Line Products
Change view
Click to view available options
Magnification Power:
1X, 4X, 10X, 40X
Catalog No. Magnification Power
S000292 1X, 4X, 10X, 40X
1 options
Catalog No. S000292 Supplier Motic Swift Line Supplier No. M3FBTI2
Add to Cart
Add to Cart
M3 - F's revolutionary technology to compare images in both micro and macro environments, included detachable 10 in. Android tablet
  • The M3-F forensic microscope offers professional features and detachable 10 in. tablet at a highly competitive price point
  • The M3's dedicated macro lens, with a large working distance, allows you to view not only bullets but also larger more bulky, evidence items
  • Swift's powerful optical system allows for images to be seen either 100% from the left microscope, 100% from the right, side-by-side, or overlapping
  • The M3-F is the first and only comparison microscope at this price level that can achieve overlapping of images
  • Combining the features of Swift's popular M3-F's advanced comparison microscope with an Android based tablet system offers unlimited possibilities
Specifications
Eyepiece WF 10X/18 mm, lockable
Head Style Trinocular
Lighting Type LED, Adjustable
Magnification Power 1X, 4X, 10X, 40X
Model M3-F-BTI2
Objective Types Achromatic
Product Line Swift Line
Product Type Trinocular Biological Microscope
Voltage 110/220 V