Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Forensic Toxicology Experiment Kit
SDPSCI_ED

Catalog No. s42879 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42879 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S42879 Supplier Edvotek™ Supplier No. 195
Only null left
Add to Cart
Add to Cart

Student's educational kit to analyze crime scene samples for presence of drugs.

  • Identify drugs, toxins in samples collected from crime scenes, victims, potential suspects
  • Uses Enzyme Linked Immunosorbent Assay (ELISA) to analyze simulated crime scene samples for the presence of drugs

Specifications

Grade 9 to Undergraduate
For Use With (Application) Used for educational purposes
Includes Instructions, simulated crime scene samples, antibodies, buffers, microtiter plates, assorted pipets, microcentrifuge tubes.
Time Range 1 Hr.
Content And Storage Refrigeration required
Class Size 10 Lab Groups
Product Type Forensic Toxicology Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.