Description
- Makes invisible bloodstains produce an intense blue chemiluminescence
- Highly sensitive bloodstain detection reagent used in crime scene investigation
- Designed for training applications, destroys DNA and is therefore not intended for forensic DNA analysis.
- Mix in a spray bottle containing 125 mL of distilled water
- Destroys DNA and prevent DNA typing of the bloodstain
- Odorless
- Fully soluble
Specifications
Specifications
| Product Type | Bloodstain Reagent |
| For Use With (Application) | Latent Bloodstain Detection |
| Quantity | 4/Pk. |