Learn More
Ted Pella Inc Forensic Field Sampler with Adhesive Carbon Tab, and clear plastic cover, 100/pk
SDP
Catalog No. NC0456489
Encompass
The Forensic Field Sampler has been designed to collect forensic evidence with minimum interference and/or contamination coming from the sampler. Comes in 3 versions: without adhesive carbon tab, with adhesive carbon tab ready to use, and covered carbon tab (plastic cover needs to be removed prior to use). Available with standard 12.7mm SEM pin mount with 8mm pin or short 12.7mm SEM pin mount with 6mm pin (ZEISS/LEO)
Catalog No. NC0456489 Supplier Ted Pella Inc Supplier No. 16255C
The Forensic Field Sampler has been designed to collect forensic evidence with minimum interference and/or contamination coming from the sampler. Comes in 3 versions: without adhesive carbon tab, with adhesive carbon tab ready to use, and covered carbon tab (plastic cover needs to be removed prior to use). Available with standard 12.7mm SEM pin mount with 8mm pin or short 12.7mm SEM pin mount with 6mm pin (ZEISS/LEO)