Learn More
Selleck Chemical LLC Foretinib S1111-5mg
SDP
Catalog No. 503108098
Encompass_Preferred
Foretinib is an ATP-competitive inhibitor of HGFR and VEGFR mostly for Met (c-Met) and KDR with IC50 of 0 4 nM and 0 9 nM in cell-free assays Less potent against Ron Flt-1/3/4 Kit (c-Kit) PDGFR / and Tie-2 and little activity to FGFR1 and EGFR Phase 2
Catalog No. 50-310-8098 Supplier Selleck Chemical LLC Supplier No. S11115MG
Add to Cart
Add to Cart
Foretinib is an ATP-competitive inhibitor of HGFR and VEGFR mostly for Met (c-Met) and KDR with IC50 of 0 4 nM and 0 9 nM in cell-free assays Less potent against Ron Flt-1/3/4 Kit (c-Kit) PDGFR / and Tie-2 and little activity to FGFR1 and EGFR Phase 2