Learn More
Medchemexpress LLC HY-10338 10mg Medchemexpress, Foretinib CAS:849217-64-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033810MG
Encompass_Preferred
Medchemexpress, HY-10338 10mg Foretinib CAS:849217-64-7 Foretinib is a multi-target tyrosine kinase inhibitor with IC50s of 0.4 nM and 0.9 nM for Met and KDR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8729