Learn More
Apexbio Technology LLC Foretinib (GSK1363089) 849217-64-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063595
Encompass_Preferred
Foretinib (GSK1363089 CAS 849217-64-7) is a small molecule inhibitor targeting receptor tyrosine kinases involved in angiogenesis and tumor progression notably receptors for vascular endothelial growth factor (VEGF) and hepatocyte growth factor (HGF) Foretinib inhibits tyrosine kinase activity of Met Ron KDR Flt-1 Flt-4 KIT Flt-3 Tie-2 and PDGFR / exhibiting IC50 values ranging from 0 4 to 9 6 nmol/L In cell-based assays foretinib reduces MET phosphorylation in mouse B16F10 melanoma and human PC-3 prostate cells (IC50 21 23 nmol/L) inhibits HGF-induced cell migration invasion and suppresses proliferation in B16F10 A549 and HT29 tumor cell lines Foretinib is utilized in preclinical cancer research targeting angiogenesis invasion and metastasis pathways
Catalog No. 50-000-63595 Supplier Apexbio Technology LLC Supplier No. A29745.1
 Restricted Item
Add to Cart
Add to Cart
Foretinib (GSK1363089 CAS 849217-64-7) is a small molecule inhibitor targeting receptor tyrosine kinases involved in angiogenesis and tumor progression notably receptors for vascular endothelial growth factor (VEGF) and hepatocyte growth factor (HGF) Foretinib inhibits tyrosine kinase activity of Met Ron KDR Flt-1 Flt-4 KIT Flt-3 Tie-2 and PDGFR / exhibiting IC50 values ranging from 0 4 to 9 6 nmol/L In cell-based assays foretinib reduces MET phosphorylation in mouse B16F10 melanoma and human PC-3 prostate cells (IC50 21 23 nmol/L) inhibits HGF-induced cell migration invasion and suppresses proliferation in B16F10 A549 and HT29 tumor cell lines Foretinib is utilized in preclinical cancer research targeting angiogenesis invasion and metastasis pathways