Learn More
Eurofins Discoverx Foretinib (GSK1363089), 5 mg. (92-1238)
Supplier:  Eurofins Discoverx 921238
Encompass
Foretinib (GSK1363089) is a synthetic small anticancer molecule. Potently and ATP-competitively inhibits MET and VEGFR2 (IC50=0.4 and 0.9 nM, respectively), kinase enzymes believed to promote tumor growth and angiogenesis. Previously underwent clinical trials for breast cancer and NSCLC but no product made it to market. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9759