Learn More
Biotium Forget-Me-Not™ Universal Probe Master Mix with ROX (500 reactions)
Catalog No. NC1629070
Encompass_Preferred
High-performance product for fluorescent probe-based PCR applications, including quantitation and SNP genotyping. Suitable for hydrolysis probes (TaqMan and dual-Labeled BHQ probes) and displacement probes. Excellent concordance of singleplex and multiplex. Standard and fast protocol compatible.
Catalog No. NC1629070 Supplier Biotium Supplier No. 310441
High-performance product for fluorescent probe-based PCR applications, including quantitation and SNP genotyping. Suitable for hydrolysis probes (TaqMan and dual-Labeled BHQ probes) and displacement probes. Excellent concordance of singleplex and multiplex. Standard and fast protocol compatible.