Learn More
Medchemexpress LLC Forigerimod trifluoroacetate | 00-00-0 | 99.9% | C117H181N34O32PS·xC2HF3O2 | 5 MG
SDP
Catalog No. 5000243241
Encompass_Preferred
Forigerimod TFA is a 21-amino-acid peptide fragment supplied as the trifluoroacetate salt. It acts as a CD4 T-cell modulator and has been used in research on autophagy and autoimmune disorders, including systemic lupus erythematosus. Supplied as a powder with high listed purity, it is intended for research use in cellular and immunology studies.
  • 21-amino-acid peptide, TFA salt
  • High purity suitable for research (listed 99.88%)
  • Soluble in DMSO and water at 100 mg/mL with sonication
  • Powder storage: -80°C (2 years) or -20°C (1 year)
  • In solvent storage: -80°C (6 months) or -20°C (1 month)
Catalog No. 50-002-43241 Supplier Medchemexpress LLC Supplier No. HY105037A5MG
Forigerimod TFA is a 21-amino-acid peptide fragment supplied as the trifluoroacetate salt. It acts as a CD4 T-cell modulator and has been used in research on autophagy and autoimmune disorders, including systemic lupus erythematosus. Supplied as a powder with high listed purity, it is intended for research use in cellular and immunology studies.
  • 21-amino-acid peptide, TFA salt
  • High purity suitable for research (listed 99.88%)
  • Soluble in DMSO and water at 100 mg/mL with sonication
  • Powder storage: -80°C (2 years) or -20°C (1 year)
  • In solvent storage: -80°C (6 months) or -20°C (1 month)