Learn More
Grainger Fork End 304 SS Cable Size 1/16
Catalog No. 502768249
Encompass_Preferred
The Loos & Co. Wire Rope Clevis Fitting is a high quality clevis made of drop forged steel with a working load limit of 1 ton. It has a clevis opening of 1 inch and is designed for use with 1/16 inch galvanized wire rope. The clevis fitting allows for adjustable connections and anchoring and features a hot dip galvanized finish for corrosion resistance. With its compact design, it provides a secure method of terminating wire rope for overhead lifting applications. This durable clevis fitting is made in the USA.
Catalog No. 50-276-8249 Supplier Grainger Supplier No. 16X649
The Loos & Co. Wire Rope Clevis Fitting is a high quality clevis made of drop forged steel with a working load limit of 1 ton. It has a clevis opening of 1 inch and is designed for use with 1/16 inch galvanized wire rope. The clevis fitting allows for adjustable connections and anchoring and features a hot dip galvanized finish for corrosion resistance. With its compact design, it provides a secure method of terminating wire rope for overhead lifting applications. This durable clevis fitting is made in the USA.