Learn More
Grainger Fork End 304 SS Cable Size 1/4
Catalog No. 502768223
Encompass_Preferred
The LOOS Fork End 1/4 in Cable Size is a high quality steel fork end with a 1/4 inch hole size. It is zinc plated for corrosion resistance and has a working load limit of 1,260 lbs. This fork end can be used for a variety of rigging and lifting applications, fitting 1/4 inch diameter wire rope. It measures 2 inches long by 1 1/4 inches wide. Made in the USA by Loos & Co., this fork end conforms to ASME B30.26 specifications for quality and performance. The fork end is sold individually.
Catalog No. 50-276-8223 Supplier Grainger Supplier No. 16X655
The LOOS Fork End 1/4 in Cable Size is a high quality steel fork end with a 1/4 inch hole size. It is zinc plated for corrosion resistance and has a working load limit of 1,260 lbs. This fork end can be used for a variety of rigging and lifting applications, fitting 1/4 inch diameter wire rope. It measures 2 inches long by 1 1/4 inches wide. Made in the USA by Loos & Co., this fork end conforms to ASME B30.26 specifications for quality and performance. The fork end is sold individually.