Learn More
Grainger Fork End 304 SS Cable Size 1/8
Catalog No. 502765633
Encompass_Preferred
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis fitting accepts 1/16" diameter wire rope and features a cotter pin hole for securing the wire rope. Overall length is 1-1/4" and weight is 0.8 oz. This clevis fitting is ideal for light duty wire rope applications.
Catalog No. 50-276-5633 Supplier Grainger Supplier No. 16X651
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis fitting accepts 1/16" diameter wire rope and features a cotter pin hole for securing the wire rope. Overall length is 1-1/4" and weight is 0.8 oz. This clevis fitting is ideal for light duty wire rope applications.