Learn More
Grainger Loos & Co. Wire Rope Clevis Fork End, 3/16 in., Stainless Steel, 1 ea.
Catalog No. 502767131
Encompass_Preferred
This fork end clevis fitting is designed for 3/16 in. wire rope applications. Manufactured from corrosion-resistant 304 stainless steel, it meets the requirements of MIL-DTL-781. The swage-style terminal provides reliable performance in high-strength tension applications.
  • Manufactured in accordance with MIL-DTL-781 and DFARS compliant
  • Breaking strength of 4,200 lbs.
  • Designed for 3/16 in. diameter wire rope
  • Features a fork style design with a 0.313 in. bolt hole diameter
  • Constructed from 304 stainless steel for corrosion resistance
Catalog No. 50-276-7131 Supplier Grainger Supplier No. 16X653
This fork end clevis fitting is designed for 3/16 in. wire rope applications. Manufactured from corrosion-resistant 304 stainless steel, it meets the requirements of MIL-DTL-781. The swage-style terminal provides reliable performance in high-strength tension applications.
  • Manufactured in accordance with MIL-DTL-781 and DFARS compliant
  • Breaking strength of 4,200 lbs.
  • Designed for 3/16 in. diameter wire rope
  • Features a fork style design with a 0.313 in. bolt hole diameter
  • Constructed from 304 stainless steel for corrosion resistance