Learn More
Grainger Fork End 304 SS Cable Size 3/32
Catalog No. 502768006
Encompass_Preferred
The Grainger 16X650 is a 16-inch diameter, 650 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with permanently lubricated ball bearings and automatic thermal overload protection. The fan has a galvanized steel housing with baked gray enamel finish and comes with wall mounting brackets. It is suitable for commercial and industrial exhaust ventilation applications. The maximum operating temperature is 104°F and it weighs approximately 31 lbs.
Catalog No. 50-276-8006 Supplier Grainger Supplier No. 16X650
The Grainger 16X650 is a 16-inch diameter, 650 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with permanently lubricated ball bearings and automatic thermal overload protection. The fan has a galvanized steel housing with baked gray enamel finish and comes with wall mounting brackets. It is suitable for commercial and industrial exhaust ventilation applications. The maximum operating temperature is 104°F and it weighs approximately 31 lbs.