Learn More
Grainger Loos & Co. Inc. Fork End Fitting, 3/8 in. Rope Diameter, Stainless Steel
Catalog No. 502767219
Encompass_Preferred
These MS20667 Fork End fittings are designed for use with wire rope, providing reliable connections for various rigging and lifting applications. Manufactured from 304 grade stainless steel, these fittings meet the requirements of MIL-DTL-781 and are fully DFARS compliant.
  • Manufactured in the USA to military specifications
  • Compatible with both stainless steel and galvanized wire rope
  • Suitable for lifting applications
  • Swage-style fitting design
  • 3/8 in. compatible cable diameter
  • 1/2 in. pin diameter
Catalog No. 50-276-7219 Supplier Grainger Supplier No. 16X658
These MS20667 Fork End fittings are designed for use with wire rope, providing reliable connections for various rigging and lifting applications. Manufactured from 304 grade stainless steel, these fittings meet the requirements of MIL-DTL-781 and are fully DFARS compliant.
  • Manufactured in the USA to military specifications
  • Compatible with both stainless steel and galvanized wire rope
  • Suitable for lifting applications
  • Swage-style fitting design
  • 3/8 in. compatible cable diameter
  • 1/2 in. pin diameter