Learn More
Grainger Fork End 304 SS Cable Size 5/16
Catalog No. 502765067
Encompass_Preferred
The Loos & Co. 3/16" Wire Rope Clevis Fitting is made from drop forged steel for durability. It has a working load limit of 1,260 lbs and is rated for general purpose use. This clevis fitting is designed to provide a termination point for 3/16" diameter 7x7 or 7x19 wire rope. It has a clevis opening of 5/8" to allow for easy installation over pins and shafts. The Loos wire rope clevis is zinc plated for corrosion resistance. It conforms to Federal Specification FF-C-450 Type I Class 1 and Military Specification MIL-C-24613. This is a high quality wire rope end fitting suitable for a variety of general purpose applications.
Catalog No. 50-276-5067 Supplier Grainger Supplier No. 16X657
The Loos & Co. 3/16" Wire Rope Clevis Fitting is made from drop forged steel for durability. It has a working load limit of 1,260 lbs and is rated for general purpose use. This clevis fitting is designed to provide a termination point for 3/16" diameter 7x7 or 7x19 wire rope. It has a clevis opening of 5/8" to allow for easy installation over pins and shafts. The Loos wire rope clevis is zinc plated for corrosion resistance. It conforms to Federal Specification FF-C-450 Type I Class 1 and Military Specification MIL-C-24613. This is a high quality wire rope end fitting suitable for a variety of general purpose applications.